Skip to Content
Merck
All Photos(1)

Key Documents

CLS3470

Corning® Transwell® 24 well plates

0.4 μm pore size, Tissue Culture (TC)-treated surface clear polystyrene plate, round wells, polyester membrane (Polyethylene Terephthalate (PET), flat bottom wells clear sterile, case of 48, pack of 12

Synonym(s):

cell culture inserts, plate inserts, tissue culture dishes, tissue culture plates

Sign Into View Organizational & Contract Pricing

Select a Size

48 EA
$153.00

$153.00


Estimated to ship onMarch 25, 2025



Select a Size

Change View
48 EA
$153.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.13
material:
clear polystyrene plate
flat bottom wells clear
polyester membrane (Polyethylene Terephthalate (PET)
round wells
pore size:
0.4 μm
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface

$153.00


Estimated to ship onMarch 25, 2025


Product Name

Corning® Transwell® polyester membrane cell culture inserts, 6.5 mm Transwell with 0.4 μm pore polyester membrane insert, TC-treated, sterile, 48/cs

material

clear polystyrene plate
flat bottom wells clear
polyester membrane (Polyethylene Terephthalate (PET)
round wells

sterility

sterile

feature

lid
skirt

packaging

pack of 12
case of 48

manufacturer/tradename

Corning 3470

insert working volume

0.1 mL

membrane diam.

6.5 mm

membrane thickness

10 μm

size

24 wells

surface area

0.33 cm2 , cell growth area

well working volume

0.6 mL

color

clear

pore size

0.4 μm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • 10μm thick transparent polyester membrane
  • Treated for optimal cell attachment
  • Packaged 12 inserts in a 24 well plate, 4 plates per case
  • Excellent visibility under phase contrast microscopy
  • Sterilized by gamma radiation
Transwell cell culture inserts are convenient, easy-to-use permeable support devices for the study of both anchorage-dependent and anchorage-independent cell lines They feature a 10 mm, thin, microscopically transparent polyester membrane that is tissue culture treated for optimal cell attachment and growth.

Provides excellent cell visibility under phase contrast microscopy and allows assessment of cell viability and monolayer formation.

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sivakumar Murugadoss et al.
Particle and fibre toxicology, 17(1), 10-10 (2020-02-27)
The terms agglomerates and aggregates are frequently used in the regulatory definition(s) of nanomaterials (NMs) and hence attract attention in view of their potential influence on health effects. However, the influence of nanoparticle (NP) agglomeration and aggregation on toxicity is
Karolien Hollanders et al.
Investigative ophthalmology & visual science, 56(2), 1335-1348 (2015-01-30)
Rho kinase (ROCK) is associated with VEGF-driven angiogenesis, as well as with inflammation and fibrosis. Therefore, the effect of AMA0428, a novel ROCK inhibitor, was studied in these processes, which highly contribute to the pathogenesis of neovascular AMD. The effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service