Skip to Content
Merck
All Photos(1)

Key Documents

CLS3450

Corning® Transwell® 6 well plates

0.4 μm pore size, Tissue Culture (TC)-treated surface clear polystyrene plate, round wells, polyester membrane (Polyethylene Terephthalate (PET), flat bottom wells clear, sterile, pack of 6, case of 24

Synonym(s):

cell culture inserts, plate inserts, tissue culture dishes, tissue culture plates

Sign Into View Organizational & Contract Pricing

Select a Size

24 EA
$175.00

$175.00


In StockDetails



Select a Size

Change View
24 EA
$175.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
clear polystyrene plate
flat bottom wells clear
polyester membrane (Polyethylene Terephthalate (PET)
round wells
pore size:
0.4 μm
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface

$175.00


In StockDetails


Product Name

Corning® Transwell® polyester membrane cell culture inserts, 24 mm Transwell with 0.4 μm pore polyester membrane insert, TC-treated, sterile, 24/cs

material

clear polystyrene plate
flat bottom wells clear
polyester membrane (Polyethylene Terephthalate (PET)
round wells

sterility

sterile

feature

lid
skirt

packaging

case of 24
pack of 6

manufacturer/tradename

Corning 3450

insert working volume

1.5 mL

membrane diam.

24 mm

membrane thickness

10 μm

size

6 wells

surface area

4.67 cm2 , cell growth area

well working volume

2.6 mL

color

clear

pore size

0.4 μm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • 10μm thick transparent polyester membrane
  • Treated for optimal cell attachment
  • Packaged 6 inserts in a 6 well plate, 4 plates per case
  • Excellent visibility under phase contrast microscopy
  • Sterilized by gamma radiation
Transwell cell culture inserts are convenient, easy-to-use permeable support devices for the study of both anchorage-dependent and anchorage-independent cell lines They feature a 10 mm, thin, microscopically transparent polyester membrane that is tissue culture treated for optimal cell attachment and growth.

Provides excellent cell visibility under phase contrast microscopy and allows assessment of cell viability and monolayer formation.

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Pellicciotta et al.
The Journal of experimental biology, 223(Pt 24) (2020-12-31)
In many organs, thousands of microscopic 'motile cilia' beat in a coordinated fashion generating fluid flow. Physiologically, these flows are important in both development and homeostasis of ciliated tissues. Combining experiments and simulations, we studied how cilia from brain tissue
En Zhou et al.
Heart and vessels, 35(7), 1025-1035 (2020-04-10)
Atrial fibrillation (AF) is one of the most prevalent arrhythmias. Myocardial sleeves of the pulmonary vein are critical in the occurrence of AF. Our study aims to investigate the effect of synthetic vascular smooth muscle cells (SMCs) on gap junction
Cong Li et al.
Ultrasound in medicine & biology, 47(3), 723-732 (2020-12-03)
According to the literature, the von Hippel-Lindau (VHL) gene has a certain correlation with ovarian cancer. In this study, we investigated the effect and mechanism of ultrasound microbubble-mediated VHL on the biological function of ovarian cancer cells. Non-targeting lipid microbubbles
Thijs Pasman et al.
Membranes, 10(11) (2020-11-11)
Polymeric membranes are widely applied in biomedical applications, including in vitro organ models. In such models, they are mostly used as supports on which cells are cultured to create functional tissue units of the desired organ. To this end, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service