Skip to Content
Merck
All Photos(1)

Key Documents

CLS3382

Corning® HTS Transwell® 96 well permeable support system, accessory plates

96 well receiver plate, TC-treated, flat bottom, sterile, w/lid, clear, 10/cs

Synonym(s):

corning transwell, transwell corning

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$68.90

$68.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
10 EA
$68.90

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear polystyrene plate
flat bottom wells clear
polystyrene
size:
96 wells

$68.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

clear polystyrene plate
flat bottom wells clear
polystyrene

sterility

sterile

feature

lid
skirt

packaging

case of 10
pack of 10

manufacturer/tradename

Corning 3382

size

96 wells

color

clear

suitability

suitable for (cell culture applications; optimal for cell attachment)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Product is an accessory for the Corning® HTS Transwell®-96 Permeable Support system
  • 96 individually isolated wells that are tc-treated (for cell attachment) and sterile
  • For use in cell growth or assay steps of an experiment
  • Used to minimize contamination when change of media is needed or to prevent cross contamination when media type is switched during cell growth step
  • When used with HTS Transwell®-96 unit, the plate allows for convenient pipette access to both apical and basolateral sides of each well (manual or automated)
  • Corner notched and SBS standard footprint
  • Sterile lid included with each plate

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service