Skip to Content
Merck
All Photos(1)

Key Documents

PHL80465

Gramine

phyproof® Reference Substance

Synonym(s):

3-(Dimethylaminomethyl)indole, Donaxine, NSC 16892

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$497.00

$497.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$497.00

About This Item

Empirical Formula (Hill Notation):
C11H14N2
CAS Number:
Molecular Weight:
174.24
Beilstein:
140521
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$497.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

mp

132-134 °C (lit.)

format

neat

SMILES string

CN(C)Cc1c[nH]c2ccccc12

InChI

1S/C11H14N2/c1-13(2)8-9-7-12-11-6-4-3-5-10(9)11/h3-7,12H,8H2,1-2H3

InChI key

OCDGBSUVYYVKQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

Reactant for preparation of:
  • Dopamine D2 receptor antagonists
  • Anti-malarial drugs
  • 5-indolyl-Mannich bases
  • Proliferation inhibitors
  • Inhibitors of human mast cell chymase
  • Preparation of DL-tryptophan
  • Potential detoxification inhibitors of the crucifer phytoalexin brassinin
  • 3-vinylindoles
  • Serotonin 5-HT6 receptor ligand templates
  • Selective protein kinase c delta (PKCδ) down regulators

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

332.6 °F

Flash Point(C)

167 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service