Skip to Content
Merck
All Photos(4)

Key Documents

C13408

Sigma-Aldrich

O-(Carboxymethyl)hydroxylamine hemihydrochloride

98%

Synonym(s):

Carboxymethoxylamine hemihydrochloride, (Aminooxy)acetic acid hemihydrochloride, (Carboxymethoxy)amine hemihydrochloride, Hydroxylamine-O-acetic acid hemihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$129.00
10 G
$406.00
25 G
$455.00

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$129.00
10 G
$406.00
25 G
$455.00

About This Item

Linear Formula:
NH2OCH2COOH · 0.5HCl
CAS Number:
Molecular Weight:
109.30
Beilstein:
3680528
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

synthetic (organic)

Quality Level

Assay

98%

form

crystalline powder
powder or crystals

mp

156 °C (dec.) (lit.)

solubility

water: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

Cl.NOCC(O)=O.NOCC(O)=O

InChI

1S/2C2H5NO3.ClH/c2*3-6-1-2(4)5;/h2*1,3H2,(H,4,5);1H

InChI key

KBXIJIPYZKPDRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

O-(Carboxymethyl)hydroxylamine hemihydrochloride can be used as a reactant to synthesize oximes via condensation with aldehydes and ketones [1]. It can also be used as a potent inhibitor of (pyridoxal 5’-phosphate) PLP-dependent β-lyases [2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of a 5'-aldehyde terminus resulting from the oxidative attack at C5 `of a 2-deoxyribose on DNA
Angeloff A, et al.
Chemical Research in Toxicology, 14, 1413-1420 (2001)
Tissue distribution of cytosolic beta-elimination reactions of selenocysteine Se-conjugates in rat and human
Rooseboom M, et al.
Chemico-Biological Interactions, 140, 243-264 (2002)
Tetrahedron Letters, 34, 4347-4347 (1993)
Seon-Ae Choi et al.
Scientific reports, 6, 26203-26203 (2016-05-18)
Hydrogen sulfide (H2S) is an important biological messenger, but few biologically-compatible methods are available for its detection in aqueous solution. Herein, we report a highly water-soluble naphthalimide-based fluorescent probe (L1), which is a highly versatile building unit that absorbs and
Ches'Nique M Phillips et al.
Cancer research, 77(21), 5741-5754 (2017-09-20)
The trans-sulfuration enzyme cystathionine-β-synthase (CBS) and its product hydrogen sulfide (H2S) are aberrantly upregulated in colorectal cancers, where they contribute to tumor growth and progression by both autocrine and paracrine mechanisms. However, it is unknown whether the CBS/H2S axis plays

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service