Skip to Content
Merck
All Photos(1)

Documents

PHR1003

Supelco

Aspirin (Acetyl Salicylic Acid)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Acetylsalicylic acid, 2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
779271
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 617
traceable to Ph. Eur. A0200000
traceable to USP 1044006

API family

aspirin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

134-136 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

BSYNRYMUTXBXSQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Aspirin (Acetyl Salicylic Acid) is a cyclo-oxygenase inhibitor and an antiplatelet agent, widely used for the secondary preventions of heart attack and stroke.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Aspirin (Acetyl Salicylic Acid) may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric technique and reversed-phase high-performance liquid chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.
Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7669 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

482.0 °F

Flash Point(C)

250 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preliminary studies on the analgesic activity of latex of Calotropris procera
Dewan.S, et al.
Journal of Ethnopharmacology, 73, 307-311 (2000)
Studies on the anti-inflammatory and analgesic activity of Cedrus deodara (Roxb.) Loud. wood oil
Shinde.AU, et al.
Journal of Ethnopharmacology, 65, 21-27 (1999)
Simultaneous determination of clopidogrel and aspirin in pharmaceutical dosage forms
Mishra P and Dolly A
Indian Journal of Pharmaceutical Sciences, 63(3) (2006)
Rapid and simultaneous determination of aspirin and dipyridamole in pharmaceutical formulations by reversed-phase high performance liquid chromatography (RP-HPLC) method
Prakash K, et al.
African Journal of Pharmacy and Pharmacology, 5(2), 244-251 (2011)
Gregg W Stone et al.
Lancet (London, England), 382(9892), 614-623 (2013-07-31)
The relation between platelet reactivity and stent thrombosis, major bleeding, and other adverse events after coronary artery implantation of drug-eluting stents has been incompletely characterised. We aimed to determine the relation between platelet reactivity during dual therapy with aspirin and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service