Skip to Content
Merck
All Photos(2)

Key Documents

G10806

Sigma-Aldrich

Gramine

97.5%

Synonym(s):

3-(Dimethylaminomethyl)indole, Donaxine, NSC 16892

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$99.40

$99.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$99.40

About This Item

Empirical Formula (Hill Notation):
C11H14N2
CAS Number:
Molecular Weight:
174.24
Beilstein:
140521
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$99.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97.5%

mp

132-134 °C (lit.)

SMILES string

CN(C)Cc1c[nH]c2ccccc12

InChI

1S/C11H14N2/c1-13(2)8-9-7-12-11-6-4-3-5-10(9)11/h3-7,12H,8H2,1-2H3

InChI key

OCDGBSUVYYVKQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Dopamine D2 receptor antagonists
  • Anti-malarial drugs
  • 5-indolyl-Mannich bases
  • Proliferation inhibitors
  • Inhibitors of human mast cell chymase
  • Preparation of DL-tryptophan
  • Potential detoxification inhibitors of the crucifer phytoalexin brassinin
  • 3-vinylindoles
  • Serotonin 5-HT6 receptor ligand templates
  • Selective protein kinase c delta (PKCδ) down regulators

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

332.6 °F

Flash Point(C)

167 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Grün et al.
Phytochemistry, 66(11), 1264-1272 (2005-05-24)
Two indole alkaloids with defense related functions are synthesized in the genus Hordeum of the Triticeae. Gramine (3(dimethyl-amino-methyl)-indole) is found in H. spontaneum and in some varieties of H. vulgare, the benzoxazinoid 2,4-dihydroxy-2H-1,4-benzoxazin-3(4H)-one (DIBOA) is detected in H. roshevitzii, H.
Timothy Niacaris et al.
The Journal of experimental biology, 206(Pt 2), 223-231 (2002-12-13)
Caenorhabditis elegans feeds by rhythmically contracting its pharynx to ingest bacteria. The rate of pharyngeal contraction is increased by serotonin and suppressed by octopamine. Using an electrophysiological assay, we show that serotonin and octopamine regulate two additional aspects of pharyngeal
Shaoyong Ke et al.
European journal of medicinal chemistry, 54, 248-254 (2012-06-01)
A series of novel gramine analogues were designed and synthesized via a convenient three-component reaction, and which were evaluated for their inhibition activities against cell proliferation. Their structures were confirmed by satisfactory spectra analyses mainly including (1)H NMR, and ESI-MS
Yu Hong et al.
Aquatic toxicology (Amsterdam, Netherlands), 91(3), 262-269 (2009-01-10)
In recent years, the exploration and development of the effective methods of treatment and prevention to algal blooms, especially Microcystis aeruginosa blooms has been an important issue in the field of water environment protection. Allelochemicals (natural plant toxins) are considered
Gabriela de la Herran et al.
Organic letters, 9(6), 961-964 (2007-02-20)
Gramine-MeI salts were useful starting materials for the synthesis of 3-benzyl- and 3-allylindoles by the 1,4-addition of boronic acids to the C=C-C=N linkages generated in situ under Rh(I)-catalysis. On the other hand, under Ir(I) catalysis, the reaction of gramines with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service