Skip to Content
Merck
All Photos(2)

Key Documents

840034C

Avanti

16:0-18:1 PS (POPS)

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phospho-L-serine (sodium salt); POPS; PS(16:0/18:1(9Z)); 110669

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$156.00
25 MG
$243.00
200 MG
$1,250.00
500 MG
$2,350.00

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$156.00
25 MG
$243.00
200 MG
$1,250.00
500 MG
$2,350.00

About This Item

Empirical Formula (Hill Notation):
C40H75NO10PNa
CAS Number:
Molecular Weight:
783.99
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (840034C-10mg)
pkg of 1 × 2.5 mL (840034C-25mg)
pkg of 2 × 4 mL (840034C-200mg)
pkg of 5 × 4 mL (840034C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840034C-10mg)
10 mg/mL (840034C-25mg)
25 mg/mL (840034C-200mg)
25 mg/mL (840034C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI key

RQYKXRYGZHMUSU-JEAFVVATSA-M

Packaging

5 mL Clear Glass Sealed Ampule (840034C-10mg)
5 mL Clear Glass Sealed Ampule (840034C-200mg)
5 mL Clear Glass Sealed Ampule (840034C-25mg)
5 mL Clear Glass Sealed Ampule (840034C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josef Melcr et al.
Journal of chemical theory and computation, 16(1), 738-748 (2019-11-26)
Phosphatidylserine (PS) lipids are important signaling molecules and the most common negatively charged lipids in eukaryotic membranes. The signaling can be often regulated by calcium, but its interactions with PS headgroups are not fully understood. Classical molecular dynamics (MD) simulations
Kari Kusler et al.
The journal of physical chemistry. B, 120(50), 12883-12889 (2016-12-14)
Phosphatidylserine (PS) has previously been found to bind Cu2+ in a ratio of 1 Cu2+ ion per 2 PS lipids to form a complex with an apparent dissociation constant that can be as low as picomolar. While the affinity of
Carina Drechsler et al.
Biophysical journal, 115(8), 1509-1517 (2018-09-30)
Lipid asymmetries between the outer and inner leaflet of the lipid bilayer exist in nearly all biological membranes. Although living cells spend great effort to adjust and maintain these asymmetries, little is known about the biophysical phenomena within asymmetric membranes
R J Perrin et al.
The Journal of biological chemistry, 275(44), 34393-34398 (2000-08-23)
alpha-Synuclein has been centrally implicated in neurodegenerative disease, and a normal function in developmental synaptic plasticity has been suggested by studies in songbirds. A variety of observations suggest the protein partitions between membrane and cytosol, a behavior apparently conferred by
Maria J Sarmento et al.
Langmuir : the ACS journal of surfaces and colloids, 33(43), 12463-12477 (2017-09-30)
Although the evidence for the presence of functionally important nanosized phosphorylated phosphoinositide (PIP)-rich domains within cellular membranes has accumulated, very limited information is available regarding the structural determinants for compartmentalization of these phospholipids. Here, we used a combination of fluorescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service