Skip to Content
Merck
All Photos(2)

Key Documents

840034P

Avanti

16:0-18:1 PS (POPS)

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phospho-L-serine (sodium salt); POPS; PS(16:0/18:1(9Z)); 110669

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$301.00
25 MG
$467.00
200 MG
$2,400.00
500 MG
$4,490.00

$301.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$301.00
25 MG
$467.00
200 MG
$2,400.00
500 MG
$4,490.00

About This Item

Empirical Formula (Hill Notation):
C40H75NO10PNa
CAS Number:
Molecular Weight:
783.99
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$301.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-L-serine (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840034P-10mg)
pkg of 2 × 100 mg (840034P-200mg)
pkg of 5 × 100 mg (840034P-500mg)
pkg of 1 × 25 mg (840034P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI key

RQYKXRYGZHMUSU-JEAFVVATSA-M

General description

Phosphoserine/phosphatidylserine (PS) is an endogenous phospholipid,[1] that is found in all polyanionic macromolecules.[2]

Application

16:0-18:1 PS (POPS) has been used:
  • in phospholipid mixture for nanodisc preparation[3]
  • in liposome preparation[4]
  • in lipid vesicle preparation for surface plasmon resonance (SPR) analysis[5]

Biochem/physiol Actions

Phosphoserine (PS) can be used as a template for the recognition of cancer antigen 125 (CA 125).[6] It is very essential for healthy nerve cell membranes and myelin.[1] Exogenous PS can be actively absorbed in humans. It has the ability to cross the blood-brain barrier. It plays a key role in the formation of short-term memory, the ability to create new memories, to focus attention, to reason and solve problems and several other cognitive activities. It actively participates in the locomotor functions, like rapid reactions and reflexes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (840034P-10mg)
5 mL Clear Glass Sealed Ampule (840034P-200mg)
5 mL Clear Glass Sealed Ampule (840034P-25mg)
5 mL Clear Glass Sealed Ampule (840034P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorophore Labeling, Nanodisc Reconstitution and Single-molecule Observation of a G Protein-coupled Receptor
Lamichhane R, et al.
Bio-protocol, 7(12) (2017)
Cellular phosphatase activity of C1-Ten/Tensin2 is controlled by Phosphatidylinositol-3, 4, 5-triphosphate binding through the C1-Ten/Tensin2 SH2 domain
Kim E, et al.
Cellular Signalling, 51, 130-138 (2018)
Josef Melcr et al.
Journal of chemical theory and computation, 16(1), 738-748 (2019-11-26)
Phosphatidylserine (PS) lipids are important signaling molecules and the most common negatively charged lipids in eukaryotic membranes. The signaling can be often regulated by calcium, but its interactions with PS headgroups are not fully understood. Classical molecular dynamics (MD) simulations
Examination of the Interaction between a Membrane Active Peptide and Artificial Bilayers by Dual Polarisation Interferometry
Payne JAE, et al.
Bio-protocol, 7(1), e2087-e2087 (2017)
Kari Kusler et al.
The journal of physical chemistry. B, 120(50), 12883-12889 (2016-12-14)
Phosphatidylserine (PS) has previously been found to bind Cu2+ in a ratio of 1 Cu2+ ion per 2 PS lipids to form a complex with an apparent dissociation constant that can be as low as picomolar. While the affinity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service