Skip to Content
Merck
All Photos(2)

Key Documents

850155P

Avanti

18:1 PI(4,5)P2

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-[phosphoinositol-4,5-bisphosphate] (ammonium salt); PIP2[4′,5′](18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$174.00
500 μG
$695.00

$174.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$174.00
500 μG
$695.00

About This Item

Empirical Formula (Hill Notation):
C45H94N3O19P3
CAS Number:
Molecular Weight:
1074.16
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$174.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-4′,5′-bisphosphate) (ammonium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850155P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850155P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](OP(O)([O-])=O)[C@H](OP([O-])(O)=O)[C@@H](O)[C@H]1O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+].[NH4+].[NH4+]

General description

Although PI(4,5)P2 is a minor component of cell membranes, it plays a critical role as a substrate for a number of important signaling proteins. PI(4,5)P2 is an intermediate in the IP3/DAG pathway where it is hydrolyzed by phospholipase C to liberate the second messengers, inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG). PI(4,5)P2 is also a substrate for PI 3-kinase where it is phosphorylated to PI(3,4,5)P3, an activator of downstream signaling components such as the protein kinase AKT.
Phosphatidylinositol is a structural phospholipid.[1] Phosphatidylinositol phosphate (PIP) is located in the plasma membrane.[2]

Application

18:1 PI(4,5)P2 is suitable to use as a supplement in native macrophage membranes to generate phospholipid enriched membrane liposome.s[3] It has also been used:
  • in the preparation of anionic phospholipid liposomes[4]
  • as a component in supported membrane tube (SMrT) templates[5]
  • in liposome preparation[6]

Biochem/physiol Actions

Phosphatidylinositol bisphosphate (PIP2) can effectively stimulate the ATPase.[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850155P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850155P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylinositol Turnover and Receptors
Ramazzotti G, et al.
Archives of Biochemistry and Biophysics (2016)
Use of the supported membrane tube assay system for real time analysis of membrane fission reactions
Dar S, et al.
Nature Protocols, 12(2), 390-390 (2017)
Dissociation of Rac1 (GDP)-RhoGDI complexes by the cooperative action of anionic liposomes containing phosphatidylinositol 3, 4, 5-trisphosphate, Rac guanine nucleotide exchange factor, and GTP
Ugolev Y, et al.
The Journal of Biological Chemistry, 283(32), 22257-22271 (2008)
Tripartite Chimeras Comprising Functional Domains Derived from the Cytosolic NADPH Oxidase Components p47phox, p67phox, and Rac1 Elicit Activator-independent Superoxide Production by Phagocyte Membranes AN ESSENTIAL ROLE FOR ANIONIC MEMBRANE PHOSPHOLIPIDS
Berdichevsky Y, et al.
The Journal of biological chemistry, 282(30), 22122- 22139 (2007)
Qualitative and quantitative characterization of protein-phosphoinositide interactions with liposome-based methods
Busse RA, et al.
Autophagy, 9(5), 770-777 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service