Skip to Content
Merck
All Photos(2)

Key Documents

840039C

Avanti

18:0-18:1 PS

1-stearoyl-2-oleoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform

Synonym(s):

1-octadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-L-serine (sodium salt); SOPS; PS(18:0/18:1(9Z)); 110672

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$234.00

$234.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$234.00

About This Item

Empirical Formula (Hill Notation):
C42H79NO10PNa
CAS Number:
Molecular Weight:
812.04
UNSPSC Code:
12352211
NACRES:
NA.25

$234.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (840039C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840039C

concentration

10 mg/mL (840039C-10mg)

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C42H80NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(44)50-35-38(36-51-54(48,49)52-37-39(43)42(46)47)53-41(45)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h18,20,38-39H,3-17,19,21-37,43H2,1-2H3,(H,46,47)(H,48,49);/q;+1/p-1/b20-18-;/t38-,39+;/m1./s1

InChI key

ALPKKMIPHGSQRX-NJZWBUMZSA-M

General description

18:0-18:1 PS (1-stearoyl-2-oleoyl-sn-glycero-3-phospho-L-serine) is a sodium salt of a modified lipid which has a phosphoserine head group. This anionic lipid[1] has asymmetric acyl chains.[2]

Application

18:0-18:1 PS (1-stearoyl-2-oleoyl-sn-glycero-3-phospho-L-serine (sodium salt)) may be used:
  • in the preparation of liposomes[3]
  • for liposome binding assays[1]
  • to generate multilamellar phospholipid vesicles[4]

Biochem/physiol Actions

Phosphatidylserine (PtdSer) plays a key role in various intracellular signaling pathways. It also participates in apoptosis and blood clotting.[5]

Packaging

5 mL Clear Glass Sealed Ampule (840039C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Zhou et al.
Cell, 168(1-2), 239-251 (2017-01-04)
K-Ras is targeted to the plasma membrane by a C-terminal membrane anchor that comprises a farnesyl-cysteine-methyl-ester and a polybasic domain. We used quantitative spatial imaging and atomistic molecular dynamics simulations to examine molecular details of K-Ras plasma membrane binding. We
Julie K Freed et al.
Endothelium : journal of endothelial cell research, 15(4), 225-230 (2008-07-30)
Previous studies have demonstrated that endothelial cells exposed to laminar shear stress are protected from apoptotic stimuli such as tumor necrosis factor (TNF)-alpha. The authors investigated the role of phosphatidylserine (PS) in this phenomenon. Western blot analysis of cleaved caspase
Jason G Kay et al.
Cell communication and signaling : CCS, 17(1), 126-126 (2019-10-17)
Phosphatidylserine (PtdSer), an essential constituent of eukaryotic membranes, is the most abundant anionic phospholipid in the eukaryotic cell accounting for up to 10% of the total cellular lipid. Much of what is known about PtdSer is the role exofacial PtdSer
Anne Jäkel et al.
Protein & cell, 1(2), 188-197 (2011-01-05)
The role of surfactant protein A (SP-A) in the recognition and clearance of apoptotic cells is well established, but to date, it is still not clear which surface molecules of apoptotic cells are involved in the process. Here we present
Marco M Manni et al.
eLife, 7 (2018-03-16)
Phospholipid membranes form cellular barriers but need to be flexible enough to divide by fission. Phospholipids generally contain a saturated fatty acid (FA) at position sn1 whereas the sn2-FA is saturated, monounsaturated or polyunsaturated. Our understanding of the impact of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service