Skip to Content
Merck
All Photos(3)

Key Documents

810158P

Avanti

16:0 Liss Rhod PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$375.00
5 MG
$598.00
10 MG
$851.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$375.00
5 MG
$598.00
10 MG
$851.00

About This Item

Empirical Formula (Hill Notation):
C64H105N4O14PS2
CAS Number:
Molecular Weight:
1249.64
UNSPSC Code:
12352211
NACRES:
NA.25

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810158P-1mg)
pkg of 5 × 1 mg (810158P-5mg)
pkg of 5 × 2 mg (810158P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNS(=O)(C1=CC(S(=O)([O-])=O)=C(C2=C3C=CC(C=C3OC4=C2C=CC(N(CC)CC)=C4)=[N+](CC)CC)C=C1)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C64H102N3O14PS2.H3N/c1-7-13-15-17-19-21-23-25-27-29-31-33-35-37-62(68)77-50-54(80-63(69)38-36-34-32-30-28-26-24-22-20-18-16-14-8-2)51-79-82(70,71)78-46-45-65-83(72,73)55-41-44-58(61(49-55)84(74,75)76)64-56-42-39-52(66(9-3)10-4)47-59(56)81-60-48-53(40-4

InChI key

GGSIOFFNWNFZNH-KINGROEASA-N

Application

16:0 Liss Rhod PE or 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt) has been used in:
  • brain polar lipid mixture for in vitro liposome tubulation assay[1]
  • in liposomes as a control to determine the stability of PTX-loaded liposomes[2]
  • the formation of liposome to understand the binding of kinesin motor protein (KIF5B) to phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2), responsible for autolysosome tabulation[3]

16:0 Liss Rhod PE may also be used in liposome flotation assay for the analysis of protein-lipid interactions[4] and in the formation of small unilamellar vesicles to determine membrane diffusion in lipid bilayers.[5]

Packaging

5 mL Amber Glass Screw Cap Vial (810158P-10mg)
5 mL Amber Glass Screw Cap Vial (810158P-1mg)
5 mL Amber Glass Screw Cap Vial (810158P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madi El-Haj et al.
Immunologic research, 67(4-5), 337-347 (2019-11-23)
The considerable variability of responses amongst subjects to disease triggers and immunotherapies is a major obstacle to designing better immune-based therapies. Therefore, development of patient-tailored precision medicine that improves the efficacy of immunomodulatory drugs is necessary. The individualized response to
Kazunori Takano et al.
The EMBO journal, 27(21), 2817-2828 (2008-10-17)
Extended Fer-CIP4 homology (EFC)/FCH-BAR (F-BAR) domains generate and bind to tubular membrane structures of defined diameters that are involved in the formation and fission of endocytotic vesicles. Formin-binding protein 17 (FBP17) and Toca-1 contain EFC/F-BAR domains and bind to neural
Wanqing Du et al.
Developmental cell, 37(4), 326-336 (2016-05-25)
Autophagic lysosome reformation (ALR) plays an important role in maintaining lysosome homeostasis. During ALR, lysosomes are reformed by recycling lysosomal components from autolysosomes. The most noticeable step of ALR is autolysosome tubulation, but it is currently unknown how the process
Hamed Hamedinasab et al.
International journal of biological macromolecules (2019-11-27)
The purpose of the present investigation was to formulate NAC (N-acetylcysteine)-loaded chitosan (CH)-coated liposome aiming at obtaining an effective formulation able to ensure a prolonged and controlled release of NAC to the lung by inhalation. Empty liposomes [(DPPG/Chol/DPPG with different
Javier Espadas et al.
Nature communications, 10(1), 5327-5327 (2019-11-24)
The endoplasmic reticulum (ER) is a continuous cell-wide membrane network. Network formation has been associated with proteins producing membrane curvature and fusion, such as reticulons and atlastin. Regulated network fragmentation, occurring in different physiological contexts, is less understood. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service