Skip to Content
Merck
All Photos(2)

Key Documents

810150P

Avanti

18:1 Liss Rhod PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$375.00
5 MG
$598.00
10 MG
$851.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$375.00
5 MG
$598.00
10 MG
$851.00

About This Item

Empirical Formula (Hill Notation):
C68H109N4O14PS2
CAS Number:
Molecular Weight:
1301.72
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$375.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810150P-1mg)
pkg of 5 × 1 mg (810150P-5mg)
pkg of 5 × 2 mg (810150P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

L-α-Phosphatidylethanolamine (PE) is a membrane phospholipid[1] and is the second most abundant phospholipid in animals, plants and yeast. It is the major lipid in bacteria.[2]

Application

18:1 Liss Rhod PE suitable to be used for labelling DOPC (1,2-dioleoyl-sn-glycero-3-phosphocholine) for visualization purpose.[3]

Biochem/physiol Actions

18:1 Liss Rhod PE is a fluorescent labelled lipid,[4] useful in labelling giant unilamellar vesicles (GUVs).[5] L-α-Phosphatidylethanolamine (PE) is essential for maintaining the structural integrity of membranes.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810150P-10mg)
5 mL Amber Glass Screw Cap Vial (810150P-1mg)
5 mL Amber Glass Screw Cap Vial (810150P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new class of synthetic retinoid antibiotics effective against bacterial persisters
Kim W, et al.
Nature, 556(7699), 103-103 (2018)
Spatiotemporal control of coacervate formation within liposomes
Deshpande S, et al.
Nature Communications, 10(1), 1800-1800 (2019)
Stabilization of liposomes against stress using polyelectrolytes: Interaction mechanisms, influence of pH, molecular weight, and polyelectrolyte structure
Rinaudo M, et al.
International Journal of Polymer Analysis and Characterization, 14(8), 667-677 (2009)
Yachong Guo et al.
ACS nano (2018-11-20)
Increasing awareness of bioeffects and toxicity of nanomaterials interacting with cells puts in focus the mechanisms by which nanomaterials can cross lipid membranes. Apart from well-discussed energy-dependent endocytosis for large objects and passive diffusion through membranes by solute molecules, other
Thuc T Le et al.
BMC cancer, 9, 42-42 (2009-02-03)
Lipid-rich tumours have been associated with increased cancer metastasis and aggressive clinical behaviours. Nonetheless, pathologists cannot classify lipid-rich tumours as a clinically distinctive form of carcinoma due to a lack of mechanistic understanding on the roles of lipids in cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service