Skip to Content
Merck
All Photos(3)

Key Documents

840032C

Avanti

Brain PS

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidylserine (Brain, Porcine) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$216.00
25 MG
$324.00
100 MG
$983.00

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$216.00
25 MG
$324.00
100 MG
$983.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (840032C-10mg)
pkg of 1 × 2.5 mL (840032C-25mg)
pkg of 1 × 4 mL (840032C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840032C-10mg)
10 mg/mL (840032C-25mg)
25 mg/mL (840032C-100mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C42H80NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(44)50-35-38(36-51-54(48,49)52-37-39(43)42(46)47)53-41(45)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h18,20,38-39H,3-17,19,21-37,43H2,1-2H3,(H,46,47)(H,48,49);/q;+1/p-1/b20-18-;/t38-,39+;/m1./s1

InChI key

ALPKKMIPHGSQRX-NJZWBUMZSA-M

General description

Phosphatidylserine (PS) is a natural phospholipid nutrient, which is located in the brain, lungs, heart, liver and skeletal muscle.[1] It is an important component of the cell membrane.[2]

Application

Brain PS has also been used in protein-lipid overlay (PLO) assay to evaluate the binding of SEIPIN to phosphatidic acid (PA).[3]

Biochem/physiol Actions

Phosphatidylserine (PS) regulates the activity of receptors, ion channels, enzymes and signaling molecules. It is implicated in monitoring membrane fluidity.[1] PS plays a vital role in apoptosis and blood clotting.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840032C-100mg)
5 mL Clear Glass Sealed Ampule (840032C-10mg)
5 mL Clear Glass Sealed Ampule (840032C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Gangal et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(22), 12394-12399 (1999-10-27)
Although the catalytic (C) subunit of cAMP-dependent protein kinase is N-myristylated, it is a soluble protein, and no physiological role has been identified for its myristyl moiety. To determine whether the interaction of the two regulatory (R) subunit isoforms (R(I)
Sensing phosphatidylserine in cellular membranes
Kay JG and Grinstein S
Sensors, 11(2) (2011)
The effects of phosphatidylserine on endocrine response to moderate intensity exercise
Starks MA, et al.
Journal of the International Society of Sports Nutrition, 5(1), 11-11 (2008)
Arupratan Das et al.
Nature cell biology, 14(3), 304-310 (2012-02-22)
Lipid asymmetry at the plasma membrane is essential for such processes as cell polarity, cytokinesis and phagocytosis. Here we find that a lipid flippase complex, composed of Lem3, Dnf1 or Dnf2, has a role in the dynamic recycling of the
Takaharu Ichimura et al.
The Journal of clinical investigation, 118(5), 1657-1668 (2008-04-17)
Following injury, the clearance of apoptotic and necrotic cells is necessary for mitigation and resolution of inflammation and tissue repair. In addition to macrophages, which are traditionally assigned to this task, neighboring epithelial cells in the affected tissue are postulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service