Skip to Content
Merck
All Photos(2)

Key Documents

810158C

Avanti

16:0 Liss Rhod PE

Avanti Research - A Croda Brand 810158C

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$375.00
5 MG
$598.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$375.00
5 MG
$598.00

About This Item

Empirical Formula (Hill Notation):
C64H105N4O14PS2
CAS Number:
Molecular Weight:
1249.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810158C-1mg)
pkg of 5 × 1 mL (810158C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810158C

concentration

1 mg/mL (810158C-1mg)
1 mg/mL (810158C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

16:0 Liss Rhod PE is a fluorescent phospholipid.[1]

Application

16:0 Liss Rhod PE has been used in the preparation of lipid-oil mixtures for fluorescent imaging[2] and as a tracer lipid in process of unilamellar vesicles production.[3] It is also suitable for fluorescent visualization of the phospholipids in tribiological investigation.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810158C-1mg)
5 mL Amber Glass Screw Cap Vial (810158C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three dimensional (temperature-tension-composition) phase map of mixed DOPC-DPPC vesicles: Two solid phases and a fluid phase coexist on three intersecting planes
Chen D, et al.
Biochimica et Biophysica Acta - Biomembranes, 1838(11), 2788-2797 (2014)
Dynamic morphologies of microscale droplet interface bilayers
Mruetusatorn P, et al.
Soft Matter, 10(15), 2530-2538 (2014)
Tribological Analysis of UHMWPE Tibial Implants in Unicompartmental Knee Replacements: From Retrieved to In Vitro Studies
Sava MM, et al.
BioTechnology: An Indian Journal, 13, 1-15 (2018)
Kazunori Takano et al.
The EMBO journal, 27(21), 2817-2828 (2008-10-17)
Extended Fer-CIP4 homology (EFC)/FCH-BAR (F-BAR) domains generate and bind to tubular membrane structures of defined diameters that are involved in the formation and fission of endocytotic vesicles. Formin-binding protein 17 (FBP17) and Toca-1 contain EFC/F-BAR domains and bind to neural
Madi El-Haj et al.
Immunologic research, 67(4-5), 337-347 (2019-11-23)
The considerable variability of responses amongst subjects to disease triggers and immunotherapies is a major obstacle to designing better immune-based therapies. Therefore, development of patient-tailored precision medicine that improves the efficacy of immunomodulatory drugs is necessary. The individualized response to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service