Key Documents
Z646202
Sigma-Aldrich
Height adjustable rack
Holds 6 x 1.5-15 mL tubes
Select a Size
1 EA
$74.20
$74.20
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$74.20
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
$74.20
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
Holds 6 x 1.5-15 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18882-0000
General description
Use the Scienceware Height-Adjustable Rack to securely hold up to six tubes or vials at any desired depth in a water bath or beaker. The rack has a flexible grip that easily retains mixed test tubes of varying weight and volume from 1.5 to 15 mL. Unlike floating racks, this rack′s adjustable height prevents submersion and sinking problems that might occur with heavier or unbalanced tubes.
The compact yet stable footprint saves bench and bath space and lifts in and out of a bath without splashing. All parts are steam autoclavable at 121ºC.
Choose from one of the most recent versions:
Certificates of Analysis (COA)