Key Documents
Z551880
Sigma-Aldrich
Aldrich® in-line oil bubbler with SafetyBarb® hose connectors
joint: ST/NS 14/20
Synonym(s):
bubblers, in-line bubblers, scientific bubblers
Select a Size
1 EA
$308.00
$308.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 30, 2025
Select a Size
Change View
1 EA
$308.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$308.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 30, 2025
Joint
joint: ST/NS14/20
capacity
10 mL
General description
For precise N2 pressure control during inert atmosphere reactions. Connect reaction vessel to in-line joint or use with a ballast bulb to keep pressure constant.
Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)