Key Documents
Z553875
Sigma-Aldrich
Aldrich® safety bubbler with SafetyBarb® hose connectors
capacity 15 mL , fill mark
Synonym(s):
bubblers, safety bubblers, scientific bubblers
Select a Size
1 EA
$380.00
$380.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$380.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$380.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
capacity
15 mL , fill mark
Related Categories
General description
Built-in flash arrester bulb prevents backflow. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)