Key Documents
Z552666
Sigma-Aldrich
Aldrich® reverse-flow bubbler with SafetyBarb® hose connectors
capacity 25 mL
Synonym(s):
bubblers, scientific bubblers
Select a Size
1 EA
$748.00
$748.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$748.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$748.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
capacity
25 mL
Related Categories
General description
Monitors gas flow into, or evolution from a system. Capacity 25 mL per chamber. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)