Key Documents
Z223220
Sigma-Aldrich
Aldrich® in-line oil bubbler
joint: ST/NS 14/20
Synonym(s):
bubblers, in-line bubblers, scientific bubblers
Select a Size
1 EA
$309.00
$309.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$309.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$309.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/20
capacity
10 mL
Related Categories
General description
For precise N2 pressure control during inert atmosphere reactions. Connect reaction vessel to in-line joint or use with a ballast bulb to keep pressure constant.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)