Key Documents
SYNF62241L
Synthware pressure equalizing addition funnel
1000 mL, joint: ST/NS 24/40, stopcock: PTFE
Synonym(s):
Synthware F62241L, dropping funnel
Select a Size
1 EA
$386.00
$386.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$386.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
$386.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F62241L
capacity
1000 mL
1000 mL
General description
With 1:5 PTFE stopcock
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)