Skip to Content
Merck
All Photos(2)

Key Documents

Z545791

Sigma-Aldrich

VerSA-Flow can and drum dispenser

High-flow pressure-delivery cap with 2 in. NPT thread

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.

Looking for similar products? Visit Product Comparison Guide

General description

For high-flow pressure-delivery of solvents from Aldrich cans and drums with tapered 2 inch NPT thread or 2 in. buttress threads. Precision machined, two-piece, PBT cap with special threading that permits the removal of the cap without disconnecting solvent and gas lines when changing containers.

  • Flow rate: approx 5 to 6 L/min @ 4 psi
  • Includes 8 ft × 3/8 in. I.D. FEP solvent pick-up tubing, adjustable depth through compression fitting on cap
  • On/Off spigot 1/4 in. I.D. gas hose connection
  • Built-in pressure-relief valve vents automatically in case of excessive pressure (10 psi max.)

Legal Information

VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service