Key Documents
Z529052
Sigma-Aldrich
Aldrich® can/drum adapters
Bottle-top adapter, GL 45
Select a Size
1 EA
$318.00
$318.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$318.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
$318.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Related Categories
General description
Chemically inert PBT adapters permit the attachment of pumps and bottle-top dispensers to Aldrich 5 gal cans or drums.
GL 45 to 2in. NPT thread. CERAMUS® to Aldrich 5 gallon container.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Ceramus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)