Skip to Content
Merck
All Photos(3)

Key Documents

Z530921

Sigma-Aldrich

Flexible PTFE adapter

top female joint: ST/NS 29/32, bottom male joint: ST/NS 29/32

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$289.00

$289.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$289.00

About This Item

UNSPSC Code:
41121815
NACRES:
NB.35

$289.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Joint

female joint: ST/NS top29/32
male joint: ST/NS bottom29/32

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Used for the transfer of powders and liquids. Can be compressed to approximately 190 mm or expanded to 250 mm. Glass joints with flexible corrugated PTFE tubing is pliable enough to create an offset adapter.

Other Notes

Picture is an example of male and female joint style.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service