Skip to Content
Merck
All Photos(1)

Key Documents

1.09996

Supelco

Adapter S 40

for the direct aspiration of solvents, S 40 bottle for 3 mm O.D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NA.03
Pricing and availability is not currently available.

fitting

S 40 bottle for 3 mm O.D

storage temp.

15-25°C

Related Categories

General description

Safe, easy and contamination free solvent handling is one of the main important topics in daily lab work. Our bottle adapter connects your S40-threaded glass bottles directly to the tubes of your instrument by 4 x 3mm openings. The use of this adapter reduces escaping solvent vapours dramatically as well as contaminations of the solvent to protect your health, the environment and the solvent quality.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service