Key Documents
Z529036
Sigma-Aldrich
Aldrich® can/drum adapters
Buttress-thread adapter
Select a Size
1 EA
$337.00
$337.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$337.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
$337.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
General description
2 in. NPT thread to 2 in. buttress-thread for Aldrich plastic cans and drums.
Chemically inert PBT adapters permit the attachment of pumps and bottle-top dispensers to Aldrich 5 gal cans or drums.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)