Skip to Content
Merck
All Photos(2)

Key Documents

906123

Sigma-Aldrich

(S,R,S)-AHPC-PEG2-butyl amine hydrochloride

Synonym(s):

(2S,4R)-1-((S)-2-(2-(2-((6-Aminohexyl)oxy)ethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide, (S,R,S)-AHPC-2-2-6-NH2 HCl salt, Crosslinker−E3 ligase ligand conjugate, Protein degrader building block for PROTAC® research, Template for synthesis of targeted protein degrader, VH032 conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$1,350.00

$1,350.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$1,350.00

About This Item

Empirical Formula (Hill Notation):
C32H49N5O6S · xHCl
Molecular Weight:
631.83 (free base basis)
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$1,350.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

ligand

VH032

Quality Level

Assay

≥95%

form

solid

reaction suitability

reactivity: carboxyl reactive
reagent type: ligand-linker conjugate

functional group

amine

storage temp.

2-8°C

SMILES string

NCCCCCCOCCOCC(N[C@H](C(N1[C@H](C(NCC2=CC=C(C3=C(C)N=CS3)C=C2)=O)C[C@@H](O)C1)=O)C(C)(C)C)=O.Cl

InChI

1S/C32H49N5O6S.ClH/c1-22-28(44-21-35-22)24-11-9-23(10-12-24)18-34-30(40)26-17-25(38)19-37(26)31(41)29(32(2,3)4)36-27(39)20-43-16-15-42-14-8-6-5-7-13-33;/h9-12,21,25-26,29,38H,5-8,13-20,33H2,1-4H3,(H,34,40)(H,36,39);1H/t25-,26+,29-;/m1./s1

InChI key

CWXLKVVDXUDRQV-DDLJGEAHSA-N

General description

Protein degrader builiding block (S,R,S)-AHPC-PEG2-butyl amine (HCl salt) enables the synthesis of molecules for targeted protein degradationand PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a von Hippel-Lindau (VHL)-recruiting ligand, a linker with both hydrophobic and hydrophilic moieties, and a pendant amine for reactivity with a carboxyl group on the target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a pendant amine, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.

Automate your VHL-PEG based PROTACs with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service