Skip to Content
Merck
All Photos(1)

Key Documents

902683

Sigma-Aldrich

BocNH-PEG5-acid

Synonym(s):

2,2-Dimethyl-4-oxo-3,8,11,14,17,20-hexaoxa-5-azadocosan-22-oic acid, Boc-NH-PEG5-CH2COOH

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,060.00

$1,060.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$1,060.00

About This Item

Empirical Formula (Hill Notation):
C17H33NO9
CAS Number:
Molecular Weight:
395.45
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

$1,060.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reaction type: Pegylations
reagent type: cross-linking reagent

functional group

Boc
amine
carboxylic acid

storage temp.

−20°C

SMILES string

OC(COCCOCCOCCOCCOCCNC(OC(C)(C)C)=O)=O

InChI

1S/C17H33NO9/c1-17(2,3)27-16(21)18-4-5-22-6-7-23-8-9-24-10-11-25-12-13-26-14-15(19)20/h4-14H2,1-3H3,(H,18,21)(H,19,20)

InChI key

OLLNIAGWMCSEJB-UHFFFAOYSA-N

Application

This heterobifunctional, PEGylated crosslinker features a carboxylic acid at one end and Boc-protected amino group at the other, which can be deprotected with mildly acidic conditions. The hydrophilic PEG linker facilitates solubility in biological applications. BocNH-PEG5-acid can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.

Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miyuki Mabuchi et al.
Bioorganic & medicinal chemistry letters, 25(16), 3373-3377 (2015-06-24)
This Letter presents an effective method for the identification of target proteins of bioactive compounds such as drugs, natural products, and intrinsic ligands, using an affinity resin. The application of a photo-labile linker to an affinity resin enabled the selective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service