Skip to Content
Merck
All Photos(2)

Key Documents

919969

Sigma-Aldrich

C5 Lenalidomide-PEG2-Butyl NH2 hydrochloride

≥95%

Synonym(s):

2-(2-((6-Aminohexyl)oxy)ethoxy)-N-(2-(2,6-dioxopiperidin-3-yl)-1-oxoisoindolin-5-yl)acetamide hydrochloride, C5 Lenalidomide conjugate, Crosslinker−E3 Ligase ligand conjugate, Protein degrader building block for PROTAC® research, Template for synthesis of targeted protein degrader

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$980.00

$980.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$980.00

About This Item

Empirical Formula (Hill Notation):
C23H32N4O6 · xHCl
Molecular Weight:
460.52 (free base basis)
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$980.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

ligand

C5 Lenalidomide

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reactivity: carboxyl reactive
reagent type: ligand-linker conjugate

functional group

amine

storage temp.

2-8°C

SMILES string

O=C1N(C2CCC(NC2=O)=O)CC3=CC(NC(COCCOCCCCCCN)=O)=CC=C31.Cl

InChI

1S/C23H32N4O6.ClH/c24-9-3-1-2-4-10-32-11-12-33-15-21(29)25-17-5-6-18-16(13-17)14-27(23(18)31)19-7-8-20(28)26-22(19)30;/h5-6,13,19H,1-4,7-12,14-15,24H2,(H,25,29)(H,26,28,30);1H

InChI key

HDOCYCRRBNJXSG-UHFFFAOYSA-N

Application

Protein degrader building block C5 Lenalidomide-PEG2-Butyl NH2 hydrochloride enables the synthesis of molecules for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a Cereblon (CRBN)-recruiting ligand with alternative exit vector, a linker with both hydrophobic and hydrophilic moieties, and a pendant amine for reactivity with an acid on the target warhead. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a terminal amine, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service