Skip to Content
Merck
All Photos(3)

Key Documents

901517

Sigma-Aldrich

(S,R,S)-AHPC-PEG2-Alkyne

≥95%

Synonym(s):

(2S,4R)-1-((S)-3,3-Dimethyl-2-(3-(2-(prop-2-yn-1-yloxy)ethoxy)propanamido)butanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide, Crosslinker−E3 Ligase ligand conjugate, Protein degrader building block for PROTAC® research, Template for synthesis of targeted protein degrader

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H40N4O6S
Molecular Weight:
584.73
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22

ligand

VH032

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: click chemistry
reagent type: ligand-linker conjugate

functional group

alkyne

storage temp.

2-8°C

SMILES string

O=C(NCC1=CC=C(C2=C(C)N=CS2)C=C1)[C@H](C[C@@H](O)C3)N3C([C@H](C(C)(C)C)NC(CCOCCOCC#C)=O)=O

InChI

1S/C30H40N4O6S/c1-6-12-39-14-15-40-13-11-25(36)33-27(30(3,4)5)29(38)34-18-23(35)16-24(34)28(37)31-17-21-7-9-22(10-8-21)26-20(2)32-19-41-26/h1,7-10,19,23-24,27,35H,11-18H2,2-5H3,(H,31,37)(H,33,36)/t23-,24+,27-/m1/s1

InChI key

LQLAETVTURDZMW-ONBPZOJHSA-N

Application

Protein degrader builiding block (S,R,S)-AHPC-PEG2-Alkyne enables the synthesis of molecules for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a Von Hippel-Lindau (VHL)-recruiting ligand and a PEGylated crosslinker with pendant alkyne for click chemistry with an azide on the target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a pendant alkyne group, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.

Automate your VHL-PEG based PROTACs with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Momar Toure et al.
Angewandte Chemie (International ed. in English), 55(6), 1966-1973 (2016-01-13)
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is
Kedra Cyrus et al.
Molecular bioSystems, 7(2), 359-364 (2010-10-06)
Conventional genetic approaches have provided a powerful tool in the study of proteins. However, these techniques often preclude selective manipulation of temporal and spatial protein functions, which is crucial for the investigation of dynamic cellular processes. To overcome these limitations
Momar Toure et al.
Angewandte Chemie (International ed. in English), 55(6), 1966-1973 (2016-01-13)
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is
Philipp M Cromm et al.
Cell chemical biology, 24(9), 1181-1190 (2017-06-27)
Traditional pharmaceutical drug discovery is almost exclusively focused on directly controlling protein activity to cure diseases. Modulators of protein activity, especially inhibitors, are developed and applied at high concentration to achieve maximal effects. Thereby, reduced bioavailability and off-target effects can

Articles

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service