909998
2-(2-((7-(tert-Butoxy)-7-oxoheptyl)oxy)ethoxy)acetic acid
≥95%
Synonym(s):
2-2-6 PROTAC® linker, Bifunctional linker, OtBu-PEG2-acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥95%
form
liquid
reaction suitability
reagent type: linker
functional group
carboxylic acid
ester
storage temp.
2-8°C
SMILES string
CC(C)(OC(CCCCCCOCCOCC(O)=O)=O)C
Application
This heterobifunctional, PEGylated crosslinker 2-(2-((7-(tert-butoxy)-7-oxoheptyl)oxy)ethoxy)acetic acid features a carboxyl group at one end and a t-butyl protected carboxylic acid at the other and exhibits both hydrophobic and hydrophilic moieties for use in biological applications. The linker can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation where the linker region is referred to as 2-2-6.
Other Notes
Legal Information
PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Angewandte Chemie (International ed. in English), 55(2), 807-810 (2015-11-26)
Proteolysis Targeting Chimera (PROTAC) technology is a rapidly emerging alternative therapeutic strategy with the potential to address many of the challenges currently faced in modern drug development programs. PROTAC technology employs small molecules that recruit target proteins for ubiquitination and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service