Skip to Content
Merck
All Photos(1)

Key Documents

804398

Sigma-Aldrich

[(TMEDA)Ni(o-tolyl)Cl]

95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$106.00

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$106.00

About This Item

Empirical Formula (Hill Notation):
C13H23ClN2Ni
CAS Number:
Molecular Weight:
301.48
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

SMILES string

CC1=CC=CC=C1[Ni]Cl.CN(C)CCN(C)C

InChI

1S/C7H7.C6H16N2.ClH.Ni/c1-7-5-3-2-4-6-7;1-7(2)5-6-8(3)4;;/h2-5H,1H3;5-6H2,1-4H3;1H;/q;;;+1/p-1

InChI key

NMLMESVZRUMFAE-UHFFFAOYSA-M

Application

Effective precatalyst for a variety of nickel-catalyzed transformations, including Suzuki-Miyaura, Buchwald-Hartwig and other cyclizations and oxidative couplings.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason D Shields et al.
Organic letters, 17(9), 2166-2169 (2015-04-18)
The synthesis and catalytic activity of [(TMEDA)Ni(o-tolyl)Cl], an air-stable, crystalline solid, is described. This complex is an effective precatalyst in a variety of nickel-catalyzed transformations. The lability of TMEDA allows a wide variety of ligands to be used, including mono-

Articles

Palladium catalysts in transition metal-catalyzed coupling reactions offer diversity, accessibility, and robustness over nickel catalysts.

Related Content

Doyle group focuses on nucleophilic fluorination using PyFluor and nickel catalysis with modular, air-stable nickel precatalyst.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service