Skip to Content
Merck
All Photos(1)

Key Documents

903019

Sigma-Aldrich

[4,4′-Dimethyl-2,2′-bipyridine]nickel(II) dichloride hydrate

≥95%

Synonym(s):

(4,4′-dmbpy)NiCl2

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$137.00

$137.00


Estimated to ship onApril 06, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$137.00

About This Item

Empirical Formula (Hill Notation):
C12H12Cl2N2Ni · xH2O
Molecular Weight:
313.84 (anhydrous basis)
UNSPSC Code:
12352101
NACRES:
NA.22

$137.00


Estimated to ship onApril 06, 2025


Request a Bulk Order

Assay

≥95%

form

powder or crystals

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

mp

>300 °C

Application

Ni-catalyst for cross-couplings of alpha-carbonyl pyridinium salts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service