Skip to Content
Merck
All Photos(2)

Key Documents

802948

Sigma-Aldrich

(dppf)Ni(o-tolyl)Cl

Synonym(s):

Buchwald Ni Catalyst

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$168.00
1 G
$510.00

$168.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$168.00
1 G
$510.00

About This Item

Empirical Formula (Hill Notation):
C41H35ClFeNiP2
CAS Number:
Molecular Weight:
739.66
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$168.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Assay

95%

Quality Level

form

powder

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

mp

173 °C

storage temp.

2-8°C

SMILES string

CC1=CC=CC=C1[Ni]Cl.[C]2(P(C3=CC=CC=C3)C4=CC=CC=C4)[C][C][C][C]2.[C]5(P(C6=CC=CC=C6)C7=CC=CC=C7)[C][C][C][C]5.[Fe]

InChI

1S/2C17H14P.C7H7.ClH.Fe.Ni/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;1-7-5-3-2-4-6-7;;;/h2*1-14H;2-5H,1H3;1H;;/q;;;;;+1/p-1

InChI key

LGLPATFHWYLMCO-UHFFFAOYSA-M

General description

(dppf)Ni(o-tolyl)Cl is an air stable reagent. Its crystal structure has been investigated by X-ray studies. (dppf)Ni(o-tolyl)Cl can be prepared from (Ph3P)2Ni(o-tolyl)Cl. It is widely employed as a precatalyst for various C-N bond formation reactions. Efficacy of (dppf)Ni(o-tolyl)Cl as a precatalyst for the amination of 4-n-butylchlorobenzene with morpholine under various optimized reaction conditions has been investigated.[1]

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel H Park et al.
Organic letters, 16(1), 220-223 (2013-11-29)
A new air-stable nickel precatalyst for C-N cross-coupling is reported. The developed catalyst system displays a greatly improved substrate scope for C-N bond formation to include both a wide range of aryl and heteroaryl electrophiles and aryl, heteroaryl, and alkylamines.

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

  1. Is it water soluble?

    1 answer
    1. This product is not tested for solubility. A literature search does not yield conclusive information regarding water solubility. Further experimentation would be necessary to determine this parameter. Please see the links below to review articles that may be helpful.

      Well-defined nickel and palladium precatalysts for cross-coupling
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC5637737/

      Exploiting the trifluoroethyl group as a precatalyst ligand in nickel-catalyzed Suzuki-type alkylations
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC6540912/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service