Skip to Content
Merck
All Photos(2)

Key Documents

335363

Sigma-Aldrich

[1,3-Bis(diphenylphosphino)propane]dichloronickel(II)

Synonym(s):

(1,3-dppp)NiCl2, 1,3-Bis(diphenylphosphino)propane nickel(II) chloride, Ni(dppp)Cl2

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$106.00
25 G
$197.00

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$106.00
25 G
$197.00

About This Item

Linear Formula:
[(C6H5)2P(CH2)3P(C6H5)2]NiCl2
CAS Number:
Molecular Weight:
542.04
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

mp

213 °C (dec.) (lit.)

SMILES string

Cl[Ni]Cl.C(CP(c1ccccc1)c2ccccc2)CP(c3ccccc3)c4ccccc4

InChI

1S/C27H26P2.2ClH.Ni/c1-5-14-24(15-6-1)28(25-16-7-2-8-17-25)22-13-23-29(26-18-9-3-10-19-26)27-20-11-4-12-21-27;;;/h1-12,14-21H,13,22-23H2;2*1H;/q;;;+2/p-2

InChI key

ZBQUMMFUJLOTQC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Synthesis of block-copolythiophenes
  • Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acids
  • Solid state metathesis polycondensation
  • Polymerization of poly(3-alkoxythiophene)s
  • Diastereoselective Nozaki-Hiyama-Kishi coupling reactions of C1-C19 fragment of macrolide antibiotic aplyronine A
  • Transfer condensation polymerization of conjugated polymers
[1,3-Bis(diphenylphosphino)propane]dichloronickel(II) can be used as a precursor to synthesize metal-fullerene coordination complexes by reduction method.[1]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Carc. 1A - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ?2 Complex of Nickel Bis (diphenylphosphanyl) propane with Fullerene
Konarev DV, et al.
European Journal of Inorganic Chemistry, 2011(6), 816-820 (2011)
Renxuan Xie et al.
Nature communications, 11(1), 893-893 (2020-02-16)
The glass transition temperature (Tg) is a key property that dictates the applicability of conjugated polymers. The Tg demarks the transition into a brittle glassy state, making its accurate prediction for conjugated polymers crucial for the design of soft, stretchable
Verónica Fernández-Stefanuto et al.
Frontiers in chemistry, 8, 208-208 (2020-04-09)
Surfactants have a great number of applications. Among these chemicals, petroleum sulfonates have been widely used due to their effectiveness in reducing interfacial tension. This is the case of sodium octylnaphthalene sulfonate which is a solid with a very low
Xiaoyu Li et al.
Nature communications, 8, 15909-15909 (2017-06-27)
Micelles formed by the self-assembly of block copolymers in selective solvents have attracted widespread attention and have uses in a wide variety of fields, whereas applications based on their electronic properties are virtually unexplored. Herein we describe studies of solution-processable
Caitlyn M Wolf et al.
Soft matter, 15(25), 5067-5083 (2019-06-12)
The molecular morphology and dynamics of conjugated polymers in the bulk solid state play a significant role in determining macroscopic charge transport properties. To understand this relationship, molecular dynamics (MD) simulations and quantum mechanical calculations are used to evaluate local

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service