Skip to Content
Merck
All Photos(2)

Key Documents

907111

Sigma-Aldrich

2,6-bis(N-pyrazolyl)pyridine nickel(II) bromide

Synonym(s):

(bpp)NiBr2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$194.00

$194.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$194.00

About This Item

Empirical Formula (Hill Notation):
C11H9Br2N5Ni
CAS Number:
Molecular Weight:
429.72
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$194.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder or crystals

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

SMILES string

[Ni+].[Br-].Br.[n]3(nccc3)c1nc(ccc1)[n]2nccc2

InChI

1S/C11H9N5.2BrH.Ni/c1-4-10(15-8-2-6-12-15)14-11(5-1)16-9-3-7-13-16;;;/h1-9H;2*1H;/q;;;+1/p-1

InChI key

ZVPBBPDZLUQKSN-UHFFFAOYSA-M

Application

Precatalyst for alkyl-alkyl cross-coupling; reductive cross-coupling of styrenyl aziridines, dialkyl ether formation, reductive amination.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Heinz et al.
Journal of the American Chemical Society, 140(6), 2292-2300 (2018-01-18)
This report describes a three-component, Ni-catalyzed reductive coupling that enables the convergent synthesis of tertiary benzhydryl amines, which are challenging to access by traditional reductive amination methodologies. The reaction makes use of iminium ions generated in situ from the condensation

Related Content

Doyle group focuses on nucleophilic fluorination using PyFluor and nickel catalysis with modular, air-stable nickel precatalyst.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service