Skip to Content
Merck
All Photos(1)

Key Documents

763047

Sigma-Aldrich

XantPhos-Pd-G2

Synonym(s):

Chloro[(4,5-bis(diphenylphosphino)-9,9-dimethylxanthene)-2-(2′-amino-1,1′-biphenyl)]palladium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$135.00
2 G
$512.00
5 G
$732.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$135.00
2 G
$512.00
5 G
$732.00

About This Item

Empirical Formula (Hill Notation):
C51H42ClNOP2Pd
CAS Number:
Molecular Weight:
888.71
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

feature

generation 2

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

188-196 (decomposition)

functional group

phosphine

SMILES string

Nc1ccccc1-c2ccccc2[Pd]Cl.CC3(C)c4cccc(P(c5ccccc5)c6ccccc6)c4Oc7c(cccc37)P(c8ccccc8)c9ccccc9

InChI

1S/C39H32OP2.C12H10N.ClH.Pd/c1-39(2)33-25-15-27-35(41(29-17-7-3-8-18-29)30-19-9-4-10-20-30)37(33)40-38-34(39)26-16-28-36(38)42(31-21-11-5-12-22-31)32-23-13-6-14-24-32;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h3-28H,1-2H3;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

XVPHXQHDHZRAJL-UHFFFAOYSA-M

Application

XantPhos-Pd-G2 is a second generation palladium precatalyst with improved reactivity in palladium-catalyzed C-N cross-coupling reactions.[1][2]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

11C?Acetylation of Amines with [11C] Methyl Iodide with Bis (cyclopentadienyldicarbonyliron) as the CO Source.
Dahl K and Nordeman P
European Journal of Organic Chemistry, 2017(38), 5785-5788 (2017)
Nanomole-scale high-throughput chemistry for the synthesis of complex molecules.
Santanilla A B, et al.
Science, 347(6217), 49-53 (2015)

Articles

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service