Skip to Content
Merck
All Photos(8)

Documents

741825

Sigma-Aldrich

XPhos Pd G2

98%

Synonym(s):

2nd Generation XPhos Precatalyst, Chloro(2-dicyclohexylphosphino-2′,4′,6′-triisopropyl-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II), X-Phos aminobiphenyl palladium chloride precatalyst, XPhos-Pd-G2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H59ClNPPd
CAS Number:
Molecular Weight:
786.80
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

feature

generation 2

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

202-210 °C

functional group

phosphine

SMILES string

Nc1ccccc1-c2ccccc2[Pd]Cl.Nc3ccccc3-c4ccccc4[Pd]Cl.CC(C)c5cc(C(C)C)c(-c6cccc(c6)P(C7CCCCC7)C8CCCCC8)c(c5)C(C)C.CC(C)c9cc(C(C)C)c(c(c9)C(C)C)-c%10ccccc%10P(C%11CCCCC%11)C%12CCCCC%12

InChI

1S/2C33H49P.2C12H10N.2ClH.2Pd/c1-23(2)27-21-31(24(3)4)33(32(22-27)25(5)6)26-14-13-19-30(20-26)34(28-15-9-7-10-16-28)29-17-11-8-12-18-29;1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28;2*13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;;;/h13-14,19-25,28-29H,7-12,15-18H2,1-6H3;13-14,19-25,27-28H,7-12,15-18H2,1-6H3;2*1-6,8-9H,13H2;2*1H;;/q;;;;;;2*+1/p-2

InChI key

HMRJFNBZAWHTGN-UHFFFAOYSA-L

General description

XPhos Pd G2 is a bulky monodentate biaryl ligand. It is a Buchwald′s 2nd generation preformed catalyst. It undergoes rapid reductive elimination to form a reactive, monoligated Pd(0) species.

Application

2nd Generation Buchwald Precatalysts

  • Widely used for Suzuki-Miyaura cross-coupling reactions
XPhos Pd G2 was employed as catalyst in the following reactions:
  • Palladium-catalyzed Suzuki-Miyaura coupling reactions of potassium organotrifluoroborates and sulfamates.
  • Suzuki-Miyaura cross-coupling reactions of sensitive aryl and heteroarylboronic acids.
  • Synthesis of potassium Boc-protected secondary aminomethyltrifluoroborates, via Suzuki-Miyaura cross-coupling reaction.
For small scale and high throughput uses, product is also available as ChemBeads (927775)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary A Molander et al.
Organic letters, 14(17), 4458-4461 (2012-08-31)
Seven potassium Boc-protected secondary aminomethyltrifluoroborates were prepared in a standardized two-step process. The Suzuki-Miyaura cross-coupling reaction was studied with this new class of nucleophiles, and a large variety of aryl and hetaryl chlorides provided the desired products in good to
Gary A Molander et al.
Organic letters, 15(10), 2534-2537 (2013-05-08)
Sulfamates were studied as the electrophilic partners in the palladium-catalyzed Suzuki-Miyaura cross-coupling reaction with potassium Boc-protected primary and secondary aminomethyltrifluoroborates. A broad range of substrates was successfully coupled to provide the desired products. Complex molecules containing a new carbon-carbon bond
Gary A Molander et al.
The Journal of organic chemistry, 78(8), 4123-4131 (2013-04-11)
A method has been developed for the Pd-catalyzed synthesis of α-(hetero)aryl esters and amides through a Suzuki-Miyaura cross-coupling reaction. This method avoids the use of strong base, does not necessitate inert or low temperature formation of reagents, and does not

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service