Skip to Content
Merck
All Photos(3)

Key Documents

763039

Sigma-Aldrich

XantPhos Pd G3

95%

Synonym(s):

[(4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene)-2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$73.00
1 G
$221.00
5 G
$913.00

$73.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$73.00
1 G
$221.00
5 G
$913.00

About This Item

Empirical Formula (Hill Notation):
C52H45NO4P2PdS
CAS Number:
Molecular Weight:
948.35
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$73.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

164-167 °C (decomposition)

functional group

phosphine

SMILES string

CS(=O)(=O)O[Pd]c1ccccc1-c2ccccc2N.CC3(C)c4cccc(P(c5ccccc5)c6ccccc6)c4Oc7c(cccc37)P(c8ccccc8)c9ccccc9

InChI

1S/C39H32OP2.C12H10N.CH4O3S.Pd/c1-39(2)33-25-15-27-35(41(29-17-7-3-8-18-29)30-19-9-4-10-20-30)37(33)40-38-34(39)26-16-28-36(38)42(31-21-11-5-12-22-31)32-23-13-6-14-24-32;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h3-28H,1-2H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

AJVXPGQYAKUTGX-UHFFFAOYSA-M

General description

XantPhos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. XantPhos Pd G3 is an excellent reagent for palladium catalyzed cross-coupling reactions. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

Application

XantPhos Pd G3 may be used in the following processes:
  • Negishi cross-coupling reaction during the synthesis of palmerolides.[1]
  • Aminocarbonylation of heteroaryl bromides with carbon monoxide (CO) in the presence of triethylamine.[2]
  • Coupling between polyglycosyl thiols and aglycon halides by C-S bond formation.[3]
For small scale and high throughput uses, product is also available as ChemBeads (932213)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoretentive Palladium-Catalyzed Arylation, Alkenylation, and Alkynylation of 1-Thiosugars and Thiols Using Aminobiphenyl Palladacycle Precatalyst at Room Temperature.
Bruneau A, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 21(23), 8375-8379 (2015)
A synthetic approach to palmerolides via Negishi cross coupling. The challenge of the C15-C16 bond formation.
Carrillo J, et al.
Tetrahedron Letters, 55(33), 4623-4627 (2014)
2-Aminobiphenyl Palladacycles: The ?Most Powerful? Precatalysts in C-C and C-Heteroatom Cross-Couplings.
Bruneau A, et al.
ACS Catalysis, 5(2), 1386-1396 (2015)

Articles

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

All contents in the foil bag are weighed, plated, packed, and sealed in a glove box under nitrogen.

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service