Skip to Content
Merck
All Photos(1)

Key Documents

794228

Sigma-Aldrich

N-XantPhos Pd G3

95%

Synonym(s):

(2′-Amino-2-biphenylyl)(methanesulfonato-κO)palladium - 4,6-bis(diphenylphosphino)-10H-phenoxazine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$237.00
1 G
$881.00

$237.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$237.00
1 G
$881.00

About This Item

Empirical Formula (Hill Notation):
C49H40N2O4P2PdS
CAS Number:
Molecular Weight:
921.29
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$237.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

O=S(O[Pd]C1=CC=CC=C1C2=CC=CC=C2N)(C)=O.C3(P(C4=CC=CC=C4)C5=CC=CC=C5)=C(OC(C(P(C6=CC=CC=C6)C7=CC=CC=C7)=CC=C8)=C8N9)C9=CC=C3

InChI

1S/C36H27NOP2.C12H10N.CH4O3S.Pd/c1-5-15-27(16-6-1)39(28-17-7-2-8-18-28)33-25-13-23-31-35(33)38-36-32(37-31)24-14-26-34(36)40(29-19-9-3-10-20-29)30-21-11-4-12-22-30;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h1-26,37H;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

RKPGNKWNXMCYQX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

Each AldrichCPR product will be packaged in one consolidated unit unless you specify multiple units at the time of order.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service