Skip to Content
Merck
All Photos(2)

Documents

763381

Sigma-Aldrich

XPhos Pd G3

98%, 1:1 MTBE adduct

Synonym(s):

(2-Dicyclohexylphosphino-2′,4′,6′-triisopropyl-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate, XPhos-G3-Palladacycle, XPhos-Pd-G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H62NO3PPdS
CAS Number:
Molecular Weight:
846.45
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

146-151 °C (decomposition)

functional group

phosphine

SMILES string

CC(C1=CC(C(C)C)=C(C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4)C(C(C)C)=C1)C.NC5=C(C=CC=C5)C6=C(C=CC=C6)[Pd]OS(C)(=O)=O

InChI

1S/C33H49P.C12H10N.CH4O3S.Pd/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h13-14,19-25,27-28H,7-12,15-18H2,1-6H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

NKKHABPORDUWIG-UHFFFAOYSA-M

General description

XPhos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. XPhos Pd G3 is an excellent reagent for Suzuki cross-coupling reactions. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

Application

XPhos Pd G3 may be used in the following reactions:
  • Cyanation reaction of heterocyclic halides.
  • Coupling of heteroaryl chlorides with polyfluoroaryl zinc reagents.
  • Coupling of 2,6-difluorophenylboronic acid with (hetero)aryl chlorides.
For small scale and high throughput uses, product is also available as ChemBeads (927783)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Aminobiphenyl Palladacycles: The ?Most Powerful? Precatalysts in C-C and C-Heteroatom Cross-Couplings.
Bruneau A, et al.
ACS Catalysis, 5(2), 1386-1396 (2015)

Articles

All of the preformed catalysts used in the kit are air and moisture stable complexes in their commercially available form.

Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.

Materials Included in your KITALYSIS-24PD-2PK High-Throughput Screening Kit

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

See All

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service