Skip to Content
Merck
All Photos(2)

Key Documents

763004

Sigma-Aldrich

CPhos Pd G3

95%

Synonym(s):

CPhos-G3-Palladacycle, CPhos-Pd-G3, [(2-Dicyclohexylphosphino-2′,6′-bis(N,N-dimethylamino) -1,1′-biphenyl)-2-(2′-amino-1,1′-biphenyl)] palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$92.50
250 MG
$225.00
500 MG
$399.00

$92.50

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

Select a Size

Change View
100 MG
$92.50
250 MG
$225.00
500 MG
$399.00

About This Item

Empirical Formula (Hill Notation):
C41H54N3O3PPdS
CAS Number:
Molecular Weight:
806.34
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$92.50

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

Quality Level

Assay

95%

form

solid

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

176-178 °C (decomposition)

functional group

phosphine

SMILES string

CS(=O)(=O)O[Pd]c1ccccc1-c2ccccc2N.CN(C)c3cccc(N(C)C)c3-c4ccccc4P(C5CCCCC5)C6CCCCC6

InChI

1S/C28H41N2P.C12H10N.CH4O3S.Pd/c1-29(2)25-19-13-20-26(30(3)4)28(25)24-18-11-12-21-27(24)31(22-14-7-5-8-15-22)23-16-9-6-10-17-23;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h11-13,18-23H,5-10,14-17H2,1-4H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

SMFSVINURNWOPR-UHFFFAOYSA-M

General description

CPhos Pd G3 is a modified G3 precatalyst (G3′). It has been synthesized by Buchwald and coworkers from second generation (G2) precatalyst by methylation of the amino group present on biphenyl backbone. It is a useful catalyst for various cross-coupling reactions. They are versatile catalysts. They have long life in solution and are readily soluble in various organic solvents. G3- precatalysts have been employed in various C-N bond forming reactions.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Aminobiphenyl Palladacycles: The ?Most Powerful? Precatalysts in C-C and C-Heteroatom Cross-Couplings.
Bruneau A, et al.
ACS Catalysis, 5(2), 1386-1396 (2015)

Articles

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

All contents in the foil bag are weighed, plated, packed, and sealed in a glove box under nitrogen.

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service