Skip to Content
Merck
All Photos(7)

Key Documents

569755

Sigma-Aldrich

Hoveyda-Grubbs Catalyst® M720

Hoveyda-Grubbs Catalyst® M720
1 of 2 reviewers received a sample product or took part in a promotion

Umicore, 97%

Synonym(s):

Hoveyda-Grubbs Catalyst® 2nd Generation, Hoveyda-Grubbs Catalyst® M72 (C627), (1,3-Bis-(2,4,6-trimethylphenyl)-2-imidazolidinylidene)dichloro(o-isopropoxyphenylmethylene)ruthenium, Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](2-isopropoxyphenylmethylene)ruthenium(II), Grubbs Catalyst® C627

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$63.40
500 MG
$109.00
2 G
$335.00
10 G
$780.00
25 G
$1,620.00

$63.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$63.40
500 MG
$109.00
2 G
$335.00
10 G
$780.00
25 G
$1,620.00

About This Item

Empirical Formula (Hill Notation):
C31H38Cl2N2ORu
CAS Number:
Molecular Weight:
626.62
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$63.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Ring-Opening Polymerization

mp

216-220 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)Oc1ccccc1C=[Ru](Cl)(Cl)=C2N(CCN2c3c(C)cc(C)cc3C)c4c(C)cc(C)cc4C

InChI

1S/C21H26N2.C10H12O.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-8(2)11-10-7-5-4-6-9(10)3;;;/h9-12H,7-8H2,1-6H3;3-8H,1-2H3;2*1H;/q;;;;+2/p-2

InChI key

ZRPFJAPZDXQHSM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

It′s a phosphine free version of Grubbs 2nd Generation Catalyst with comparable reactivity, but initiates more readily at lower temperatures. Efficient for metathesis of electron-deficient substrates including fluorinated olefins. Demonstrated in multiple large-scale pharmaceutical applications.
It can be used as a catalyst for olefin cross-metathesis with fluorinated olefins (CM),[1] ring-closing metathesis (RCM),[2] ring-opening metathesis (ROM),[3] and a sequence of a metathesis reaction and subsequent dihydroxylation of the newly formed double bond.[4]

Learn more about our metathesis catalysts

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure activity relationships of 5-, 6-, and 7-methyl-substituted azepan-3-one cathepsin K inhibitors.
Yamashita DS, et al.
Journal of Medicinal Chemistry, 49(5), 1597-1612 (2006)
Synthesis of the HCV protease inhibitor Vaniprevir (MK-7009) using ring-closing metathesis strategy.
Kong J, et al.
The Journal of Organic Chemistry, 77(8), 3820-3828 (2012)
S Imhof et al.
Chemical communications (Cambridge, England), (17), 1692-1693 (2002-09-21)
The E-selective cross metathesis (CM) of fluorinated olefins with various functionalised alkenes in good to excellent yields is reported.
Brian H White et al.
Journal of the American Chemical Society, 125(48), 14901-14904 (2003-12-04)
Ring-opening/ring-closing metathesis on cyclobutene-containing substrates with angular oxygen functionality provides a stereospecific introduction of 1,5-bis-dienes required for an anion-accelerated oxy-Cope rearrangement. The reaction sequence offers generally a stereocontrolled preparation of a variety of medium ring-containing bicyclic ring systems, while rearrangement
Kevin M Kuhn et al.
Organic letters, 12(5), 984-987 (2010-02-10)
A series of ruthenium catalysts have been screened under ring-closing metathesis (RCM) conditions to produce five-, six-, and seven-membered carbamate-protected cyclic amines. Many of these catalysts demonstrated excellent RCM activity and yields with as low as 500 ppm catalyst loadings.

Related Content

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

1 of 2 reviewers received a sample product or took part in a promotion

Active Filters

  1. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service