Skip to Content
Merck
All Photos(2)

Key Documents

775258

Sigma-Aldrich

Grubbs Catalyst® M202

Umicore

Synonym(s):

Grubbs Catalyst® M2, Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](3-phenyl-1H-inden-1-ylidene)(tricyclohexylphosphine)ruthenium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$102.00
500 MG
$215.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$102.00
500 MG
$215.00

About This Item

Empirical Formula (Hill Notation):
C54H69Cl2N2PRu
CAS Number:
Molecular Weight:
949.09
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$102.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

159 (d)-162 °C

storage temp.

2-8°C

SMILES string

[Ru+2](=C7N(CCN7c9c(cc(cc9C)C)C)c8c(cc(cc8C)C)C)=C4c5c(cccc5)C(=C4)c6ccccc6.P(C3CCCCC3)(C2CCCCC2)C1CCCCC1.[Cl-].[Cl-]

InChI

1S/C21H26N2.C18H33P.C15H10.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-6-12(7-3-1)15-11-10-13-8-4-5-9-14(13)15;;;/h9-12H,7-8H2,1-6H3;16-18H,1-15H2;1-9,11H;2*1H;/q;;;;;+2/p-2

InChI key

MCNUJWCJGIGFBS-UHFFFAOYSA-L

Application

  • Oxa-Cages Synthesis: Grubbs Catalyst® M202 is utilized in a synthetic approach for creating diverse oxa-cages via olefin metathesis, enhancing the scope of organic synthesis with high selectivity and efficiency in forming complex cyclic structures (Kotha et al., 2024).
  • Phosphoproline Analogues: This article details the stereocontrolled synthesis of complex bicyclic systems, a type of chemical transformation where Grubbs Catalyst® M202 could potentially be applied for enhancing reaction pathways and outcomes in pharmaceutical synthesis (Hernández-Benítez et al., 2024).

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service