Skip to Content
Merck
All Photos(2)

Documents

775398

Sigma-Aldrich

Hoveyda-Grubbs Catalyst® M710

Umicore, 97%

Synonym(s):

Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene][(2-isopropoxy)(5-trifluoroacetamido)benzylidene]ruthenium(II), Hoveyda-Grubbs Catalyst® M71

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H38Cl2F3N3O2Ru
CAS Number:
Molecular Weight:
737.64
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

storage temp.

2-8°C

SMILES string

CC(C)OC1=C(C=C(NC(C(F)(F)F)=O)C=C1)C=[Ru](Cl)(Cl)=C2N(C3=C(C)C=C(C)C=C3C)CCN2C4=C(C)C=C(C)C=C4C

InChI

1S/C21H26N2.C12H12F3NO2.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-7(2)18-10-5-4-9(6-8(10)3)16-11(17)12(13,14)15;;;/h9-12H,7-8H2,1-6H3;3-7H,1-2H3,(H,16,17);2*1H;/q;;;;+2/p-2

InChI key

TUXRUOWNSPNNTK-UHFFFAOYSA-L

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service