Skip to Content
Merck
Hoveyda-Grubbs Catalyst® M720 Umicore, 97%

Hoveyda-Grubbs Catalyst® M720

Hoveyda-Grubbs Catalyst® M720 Umicore, 97%
Synonym(s):
(1,3-Bis-(2,4,6-trimethylphenyl)-2-imidazolidinylidene)dichloro(o-isopropoxyphenylmethylene)ruthenium, Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](2-isopropoxyphenylmethylene)ruthenium(II), Grubbs Catalyst® C627
Empirical Formula (Hill Notation):
C31H38Cl2N2ORu
CAS Number:
Molecular Weight:
626.62

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing