Skip to Content
Merck
All Photos(5)

Key Documents

771082

Sigma-Aldrich

Hoveyda-Grubbs Catalyst® M2001

Umicore

Synonym(s):

Grubbs Catalyst® Z-Selective, Hoveyda-Grubbs Catalyst® MZ1c (C633), Grubbs Catalyst® C633, Grubbs Z-Selective Metathesis Catalyst, Grubbs′ Z-Selective Catalyst, [2-(1-Methylethoxy-O)phenylmethyl-C](nitrato-O,O′){rel-(2R,5R,7R)-adamantane-2,1-diyl[3-(2,4,6-trimethylphenyl)-1-imidazolidinyl-2-y lidene]}ruthenium, [2-(1-Methylethoxy-O)phenylmethyl-C](nitrato-O,O′){rel-(2R,5R,7S)-tricyclo[3.3.1.13,7]decane-2,1-diyl[3-(2,4,6-trimethylphenyl)-1-imidazolidinyl-2-ylidene]}ruthenium

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$173.00
500 MG
$756.00
2 G
$2,290.00
5 G
$3,380.00

$173.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$173.00
500 MG
$756.00
2 G
$2,290.00
5 G
$3,380.00

About This Item

Empirical Formula (Hill Notation):
C32H41N3O4Ru
CAS Number:
Molecular Weight:
632.76
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$173.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Olefin Metathesis

mp

200-215 °C

storage temp.

2-8°C

Application

Olefin Metathesis Catalyst for cis-selective ring-opening metathesis polymerization and Z-Selective Olefin Metathesis.

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin K Keitz et al.
Journal of the American Chemical Society, 134(4), 2040-2043 (2012-01-14)
Cis-selective ring-opening metathesis polymerization of several monocyclic alkenes as well as norbornene and oxanorbornene-type monomers using a C-H activated, ruthenium-based metathesis catalyst is reported. The cis content of the isolated polymers depended heavily on the monomer structure and temperature. A
Benjamin K Keitz et al.
Journal of the American Chemical Society, 134(1), 693-699 (2011-11-22)
Several new C-H-activated ruthenium catalysts for Z-selective olefin metathesis have been synthesized. Both the carboxylate ligand and the aryl group of the N-heterocyclic carbene have been altered and the resulting catalysts evaluated using a range of metathesis reactions. Substitution of

Related Content

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service