Skip to Content
Merck
All Photos(1)

Key Documents

306649

Sigma-Aldrich

tert-Butyl ethyl malonate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CCH2CO2C2H5
CAS Number:
Molecular Weight:
188.22
Beilstein:
1776681
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

83-85 °C/8 mmHg (lit.)

density

0.994 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)CC(=O)OC(C)(C)C

InChI

1S/C9H16O4/c1-5-12-7(10)6-8(11)13-9(2,3)4/h5-6H2,1-4H3

InChI key

OCOBFMZGRJOSOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

tert-Butyl ethyl malonate was used in the synthesis of polar ester-functionalized aliphatic polysulfone with remarkable thermal stability[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A remarkably thermally stable, polar aliphatic polysulfone.
Schmidt-Winkel P and Wudl F.
Macromolecules, 31(9), 2911-2917 (1998)
Xiaoqing Ren et al.
Cell reports, 26(13), 3643-3656 (2019-03-28)
CBX4, a component of polycomb repressive complex 1 (PRC1), plays important roles in the maintenance of cell identity and organ development through gene silencing. However, whether CBX4 regulates human stem cell homeostasis remains unclear. Here, we demonstrate that CBX4 counteracts
Katherine I Zhou et al.
Molecular cell, 76(1), 70-81 (2019-08-26)
N6-methyladenosine (m6A) modification occurs co-transcriptionally and impacts pre-mRNA processing; however, the mechanism of co-transcriptional m6A-dependent alternative splicing regulation is still poorly understood. Heterogeneous nuclear ribonucleoprotein G (hnRNPG) is an m6A reader protein that binds RNA through RRM and Arg-Gly-Gly (RGG)
Junjie Tony Hua et al.
Cell, 174(3), 564-575 (2018-07-24)
The prostate cancer (PCa) risk-associated SNP rs11672691 is positively associated with aggressive disease at diagnosis. We showed that rs11672691 maps to the promoter of a short isoform of long noncoding RNA PCAT19 (PCAT19-short), which is in the third intron of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service