Skip to Content
Merck
All Photos(1)

Key Documents

163872

Sigma-Aldrich

Ethyl malonyl chloride

technical grade

Synonym(s):

Ethyl (chloroformyl)acetate, Ethyl 3-chloro-3-oxopropionate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$119.00
25 G
$520.00

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$119.00
25 G
$520.00

About This Item

Linear Formula:
CH3CH2OCOCH2COCl
CAS Number:
Molecular Weight:
150.56
Beilstein:
636215
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

refractive index

n20/D 1.429 (lit.)

bp

79-80 °C/25 mmHg (lit.)

density

1.176 g/mL at 25 °C (lit.)

functional group

acyl chloride
ester

storage temp.

2-8°C

SMILES string

CCOC(=O)CC(Cl)=O

InChI

1S/C5H7ClO3/c1-2-9-5(8)3-4(6)7/h2-3H2,1H3

InChI key

KWFADUNOPOSMIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl malonyl chloride is a versatile acylating agent for propargyl alcohols[1], hydrazines[2] and amines[3].

Application

Ethyl malonyl chloride was used in the synthesis of:
  • liquid-crystalline methanofullerodendrimers[4]
  • 3,5-disubstituted 1,2,4-oxadiazole derivatives, which are potential peptidomimetic building blocks[5]
  • 3-pyrrolin-2-ones via amidation with propargylamines and subsequent base catalyzed 5-exo-dig cyclization[6]
Versatile acylating agent for propargyl alcohols,[1] hydrazines,[2] and amines.[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sudarat Yenjai et al.
Journal of photochemistry and photobiology. B, Biology, 186, 23-30 (2018-07-11)
Rational design of photoreagents with systematic modifications of their structures can provide valuable information for a better understanding of the protein photocleavage mechanism by these reagents. Variation of the length of the linker connecting the photoactive moiety with the protein
Liquid-crystalline methanofullerodendrimers which display columnar mesomorphism.
Maringa N, et al.
Journal of Materials Chemistry, 18(13), 1524-1534 (2008)
Synthesis of 3, 5-disubstituted 1, 2, 4-oxadiazoles as peptidomimetic building blocks.
Jakopin Z, et al.
Tetrahedron Letters, 48(8), 1465-1468 (2007)
Synthesis, 2019-2019 (2006)
J T Gau et al.
Cancer research, 57(17), 3830-3834 (1997-09-01)
Prostate-specific antigen (PSA) has been demonstrated to release the active form of insulin-like growth factor I in vitro (P. Cohen et al., J. Clin. Endocrinol. & Metab., 75: 1046-1053, 1992; P. Cohen et al., J. Clin. Endocrinol. & Metab., 79:

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service