Skip to Content
Merck
All Photos(1)

Documents

D97754

Sigma-Aldrich

Diethyl malonate

ReagentPlus®, 99%

Synonym(s):

Malonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOC2H5)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
774687
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.52 (vs air)

vapor pressure

1 mmHg ( 40 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.413 (lit.)

bp

199 °C (lit.)

mp

−51-−50 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)OCC

InChI

1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3

InChI key

IYXGSMUGOJNHAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl malonate is diethyl ester of malonic acid. It is widely used as a versatile building block for introducing the malonate functional group into molecules. Acylation of diethyl malonate using magnesium chloride and triethylamine is reported. K2CO3-catalyzed 1,4-addition reaction of diethyl malonate with various substituted 1,2-allenic ketones yields polyfunctionalized β,γ-unsaturated enones.

Application

Diethyl malonate was used to investigate Knoevenagel condensation reactions in microreactor using zeolite catalysts obtained by grafting amino groups onto NaX and CsNaX zeolites. It may be used in the synthesis of diethyl benzylidenemalonate by base catalyzed Knoevenagel condensation with benzaldehyde. It may be used in the synthesis of α-aryl malonates.

Packaging

Packaged in glass bottles

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengming Ma et al.
The Journal of organic chemistry, 68(23), 8996-9002 (2003-11-08)
The K2CO3 (10 mol %)-catalyzed 1,4-addition reaction of diethyl malonate with various substituted 1,2-allenic ketones leading to polyfunctionalized beta,gamma-unsaturated enones 3 or 4 was studied. With 3-unsubstituted 1-substituted-1,2-allenyl ketones, the highly selective formation of beta,gamma-unsaturated enones 4 was observed; with
Solvent-free organocatalytic Michael addition of diethyl malonate to nitroalkenes: the practical synthesis of Pregabalin and ?-nitrobutyric acid derivatives
J Liu, et al.
Tetrahedron, 67, 636-640 (2011)
Edward J Hennessy et al.
Organic letters, 4(2), 269-272 (2002-02-14)
[reaction: see text] A general method for the synthesis of alpha-aryl malonates is described. The coupling of an aryl iodide and diethyl malonate in the presence of Cs(2)CO(3) and catalytic amounts of copper(I) iodide and 2-phenylphenol affords the alpha-aryl malonate
Integration of heterogeneous catalysts into complex synthetic routes: sequential vs. one-pot reactions in a (Knoevenagel+ Mukaiyama-Michael+ hydrogenation+ transesterification) sequence.
Fraile JM, et al.
Catalysis Science & Technology, 3(2), 436-443 (2013)
Lian Jin Liu et al.
Nucleosides, nucleotides & nucleic acids, 30(10), 784-797 (2011-10-05)
Novel 5'-norcarbocyclic adenosine phosphonic acid analogues with 6'-electropositive moiety such as spirocyclopropane were designed and synthesized from the commercially available diethylmalonate 5. Regioselective Mitsunobu reaction proceeded in the presence of an allylic functional group at a low reaction temperature in

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service